Assay Details
| Gene Symbol: Rrp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145620.4 | NP_663595.1 | 8 - 9 | 772 | 75 | |
| GenBank mRNA | AF368232.1 | - | 8 - 9 | 755 | 75 | |
| AK076113.1 | - | 8 - 9 | 774 | 75 | ||
| AK156199.1 | - | 7 - 8 | 846 | 75 | ||
| BC014703.1 | - | 8 - 9 | 772 | 4237895 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrp9
Genomic Map