Assay Details
| Gene Symbol: Mmrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153127.3 | NP_694767.3 | 2 - 3 | 404 | 91 | |
| GenBank mRNA | AK077884.1 | - | 2 - 3 | 406 | 91 | |
| AK078470.1 | - | 2 - 3 | 380 | 91 | ||
| AK134285.1 | - | 2 - 3 | 444 | 91 | ||
| BC034871.1 | - | 2 - 3 | 346 | 91 | ||
| BC145845.1 | - | 2 - 3 | 324 | 40131148 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmrn2
Genomic Map