Assay Details
| Gene Symbol: Mmrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153127.3 | NP_694767.3 | 6 - 7 | 2558 | 75 | |
| GenBank mRNA | AK077884.1 | - | 6 - 7 | 2559 | 75 | |
| AK078470.1 | - | 6 - 7 | 2534 | 75 | ||
| AK134285.1 | - | 6 - 7 | 2597 | 75 | ||
| BC034871.1 | - | 6 - 7 | 922 | 75 | ||
| BC145845.1 | - | 6 - 7 | 2478 | 40131148 | 75 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414396 | BC034871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414396 | AK078470.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414396 | AK134285.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414396 | AK077884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414396 | NM_153127.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414396 | BC034871.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414396 | AK078470.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414396 | AK134285.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414396 | AK077884.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P414396 | NM_153127.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869605 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405486 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754985 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898985 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 504294 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500986 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 728382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927144 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717162 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384469 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14154 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136833 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157702 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414396 | BC034871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414396 | AK078470.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414396 | AK134285.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414396 | AK077884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414396 | NM_153127.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414396 | BC034871.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414396 | AK078470.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414396 | AK134285.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414396 | AK077884.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P414396 | NM_153127.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529663 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29529646 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02830783 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136833 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157702 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08080361 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08080356 | - | - |
| Whole Mouse Genome G4122A | A_51_P414396 | BC034871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414396 | AK078470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414396 | AK134285.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414396 | AK077884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414396 | NM_153127.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414396 | BC034871.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414396 | AK078470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414396 | AK134285.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414396 | AK077884.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P414396 | NM_153127.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107937 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08080356 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136833 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157702 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11408065 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11408079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29529745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11408083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11200804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11408074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11200813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11408070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11200812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11200799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29529661 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10414107 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414106 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414105 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414112 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414108 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414111 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439518_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456768_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437123_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420276_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10414102 | - | - |
| Mouse Genome 430 2.0 Array | 1437123_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456768_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456768_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1420276_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1439518_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439518_at | - | - |
| Mouse Genome 430 2.0 Array | 1420276_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437123_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105450_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105450_at | - | - |
| Mouse Expression Set 430 | 1439518_PM_at | - | - |
| Mouse Expression Set 430 | 1437123_PM_at | - | - |
| Mouse Expression Set 430 | 1420276_x_at | - | - |
| Mouse Expression Set 430 | 1437123_at | - | - |
| Mouse Expression Set 430 | 1456768_a_at | - | - |
| Mouse Expression Set 430 | 1439518_at | - | - |
| Mouse Expression Set 430 | 1456768_PM_a_at | - | - |
| Mouse Expression Set 430 | 1420276_PM_x_at | - | - |
| Murine Genome U74v2 Set | 171323_i_at | - | - |
| Murine Genome U74v2 Set | 130951_at | - | - |
| Murine Genome U74v2 Set | 130951_PM_at | - | - |
| Murine Genome U74v2 Set | 171323_PM_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1420276_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1420276_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456768_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439518_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456768_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437123_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437123_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439518_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 338329 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102850408 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102850408 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1740047 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001740047 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2761918 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2761918 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2494159 | - | - |
| mouse-6 v1.1 (Illumina) | scl00105450.2_179-S | - | - |
| mouse-6 v1.1 (Illumina) | scl46452.7_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1740047 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102850408 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00105450.2_179-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102850408 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740047 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2761918 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2761918 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00105450.2_179-S | - | - |

Mmrn2
Genomic Map