Assay Details
| Gene Symbol: Cd300a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170758.3 | NP_739564.1 | 6 - 7 | 1124 | 72 | |
| GenBank mRNA | AB065156.1 | - | 6 - 7 | 843 | 72 | |
| AB091765.1 | - | 6 - 7 | 850 | 72 | ||
| AB091766.1 | - | 6 - 7 | 846 | 72 | ||
| AB095675.1 | - | 6 - 7 | 947 | 72 | ||
| AK045869.1 | - | 6 - 7 | 1124 | 72 | ||
| AK053160.1 | - | 6 - 7 | 800 | 72 | ||
| AY457054.1 | - | 6 - 7 | 800 | 72 | ||
| BC139296.1 | - | 6 - 7 | 1060 | 8862318 | 72 | |
| BC139297.1 | - | 6 - 7 | 1060 | 8862319 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
serotonin secretion by mast cell
receptor-mediated endocytosis
signal transduction
negative regulation of serotonin secretion
negative regulation of B cell proliferation
positive regulation of phosphoprotein phosphatase activity
negative regulation of mast cell activation involved in immune response
negative regulation of mast cell degranulation
negative regulation of MAP kinase activity
negative regulation of fibroblast proliferation
regulation of T cell receptor signaling pathway
negative regulation of B cell receptor signaling pathway
negative regulation of NK T cell activation
negative regulation of phagocytosis, engulfment
negative regulation of neutrophil activation
negative regulation of eosinophil activation
negative regulation of activation of JAK2 kinase activity
negative regulation of eosinophil migration
phosphatidylserine binding
protein binding
phosphatidylethanolamine binding
phosphatase binding
signaling receptor activity
immune system process
serotonin secretion by mast cell
receptor-mediated endocytosis
signal transduction
negative regulation of serotonin secretion
negative regulation of B cell proliferation
positive regulation of phosphoprotein phosphatase activity
negative regulation of mast cell activation involved in immune response
negative regulation of mast cell degranulation
negative regulation of MAP kinase activity
negative regulation of fibroblast proliferation
regulation of T cell receptor signaling pathway
negative regulation of B cell receptor signaling pathway
negative regulation of NK T cell activation
negative regulation of phagocytosis, engulfment
negative regulation of neutrophil activation
negative regulation of eosinophil activation
negative regulation of activation of JAK2 kinase activity
negative regulation of eosinophil migration
phosphatidylserine binding
protein binding
phosphatidylethanolamine binding
phosphatase binding
signaling receptor activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008297 | AK045869.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008297 | NM_170758.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 392937 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524811 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624490 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758406 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543103 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246402 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876914 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131470 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107064 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168009 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141292 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008297 | AK045869.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008297 | NM_170758.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150326 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28150335 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107064 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141292 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168009 | - | - |
| Whole Mouse Genome G4122A | A_52_P492142 | BC139297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P492142 | AY457054.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P492142 | BC139296.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P492142 | AB091765.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P492142 | AB091766.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147003 | AK045869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147003 | NM_170758.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126927 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125501 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107064 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168009 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141292 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09587519 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09587527 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09587520 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09587532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28150450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09554529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09704598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09195215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09587529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09122793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09587532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09420896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09543738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09477239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28150334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09122794 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10382446 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382440 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382444 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382443 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382448 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382441 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382445 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382439 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382442 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382447 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445292_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435903_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10382438 | - | - |
| Mouse Genome 430 2.0 Array | 1445292_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435903_at | - | - |
| Mouse Genome 430 2.0 Array | 1445292_at | - | - |
| Mouse Genome 430 2.0 Array | 1435903_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217303_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217303_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100017647.mm.1 | NM_170758.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017647.mm.1 | NM_170758.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017647.mm.1 | TC1100001882.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100017648.mm.1 | NM_170758.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017648.mm.1 | NM_170758.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100017648.mm.1 | TC1100001882.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435903_PM_at | - | - |
| Mouse Expression Set 430 | 1435903_at | - | - |
| Mouse Expression Set 430 | 1445292_PM_at | - | - |
| Mouse Expression Set 430 | 1445292_at | - | - |
| Murine Genome U74v2 Set | 104023_at | - | - |
| Murine Genome U74v2 Set | 104023_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435903_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435903_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435903_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435903_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445292_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445292_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 103990142 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6760128 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006760128 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218797 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218797 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247685 | - | - |
| mouse-6 v1.1 (Illumina) | scl40740.8_132-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103990142 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6760128 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40740.8_132-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103990142 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760128 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2887986 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2887983 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2887986 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2887983 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247685 | - | - |

Cd300a
Genomic Map