Assay Details
| Gene Symbol: Has1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008215.2 | NP_032241.1 | 4 - 5 | 1112 | 72 | |
| GenBank mRNA | AK138203.1 | - | 3 - 4 | 398 | 72 | |
| BC125424.1 | - | 4 - 5 | 1091 | 40129939 | 72 | |
| BC125426.1 | - | 4 - 5 | 1091 | 40129941 | 72 | |
| D82964.1 | - | 4 - 5 | 1118 | 72 | ||
More Information
Set Membership: |
|

Has1
Genomic Map