Assay Details
| Gene Symbol: Hcst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF122905.1 | - | 1 - 2 | 39 | 63 | |
| BC069220.1 | - | 95 | 640698 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hcst
Genomic Map