Assay Details
| Gene Symbol: Hhip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020259.4 | NP_064655.4 | 3 - 4 | 1122 | 71 | |
| GenBank mRNA | AF116865.1 | - | 3 - 4 | 1103 | 71 | |
| AK031539.1 | - | 3 - 4 | 1121 | 71 | ||
| BC053012.1 | - | 3 - 4 | 1107 | 6402422 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hhip
Genomic Map