Assay Details
| Gene Symbol: Aadac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023383.1 | NP_075872.1 | 1 - 2 | 158 | 132 | |
| GenBank mRNA | AK008244.1 | - | 1 - 2 | 159 | 132 | |
| BC019999.1 | - | 1 - 2 | 140 | 4162194 | 132 | |
| BC054823.1 | - | 1 - 2 | 147 | 4162038 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394115 | NM_023383.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394115 | AK008244.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394115 | NM_023383.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P394115 | AK008244.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79607 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495725 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185788 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35958 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87752 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179171 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394115 | NM_023383.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394115 | AK008244.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394115 | NM_023383.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P394115 | AK008244.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P394115 | NM_023383.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P394115 | AK008244.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P394115 | NM_023383.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P394115 | AK008244.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175669 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22175677 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04922995 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179171 | - | - |
| Whole Mouse Genome G4122A | A_51_P394115 | NM_023383.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394115 | AK008244.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394115 | NM_023383.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P394115 | AK008244.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125689 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179171 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02419073 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02419074 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02419061 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02419064 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02419076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22175731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02419066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02927921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02927922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02765545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02765550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03111309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02292143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02419053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02419072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02765556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02419076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22175705 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10492303 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10492305 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10492302 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10492301 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10492304 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448813_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10492300 | - | - |
| Mouse Genome 430 2.0 Array | 1448813_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448813_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67758_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67758_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300003329.mm.1 | NM_023383.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300003329.mm.1 | NM_023383.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300003329.mm.1 | TC0300000482.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300003330.mm.1 | NM_023383.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300003330.mm.1 | NM_023383.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300003330.mm.1 | TC0300000482.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300003331.mm.1 | NM_023383.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300003331.mm.1 | NM_023383.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300003331.mm.1 | TC0300000482.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448813_at | - | - |
| Mouse Expression Set 430 | 1448813_PM_at | - | - |
| Murine Genome U74v2 Set | 95439_PM_at | - | - |
| Murine Genome U74v2 Set | 95439_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448813_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448813_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448813_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448813_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 823500 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6590632 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6590632 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006590632 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2728813 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2728813 | - | - |
| mouse-6 v1.1 (Illumina) | scl23150.4.1_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6590632 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23150.4.1_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590632 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2728813 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2728813 | - | - |

Aadac
Genomic Map