| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700028150.mm.1 |
NM_001256130.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028150.mm.1 |
NM_001256130.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028150.mm.1 |
NM_001256131.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028150.mm.1 |
NM_001256131.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028150.mm.1 |
NM_001256132.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028150.mm.1 |
NM_001256132.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028150.mm.1 |
NM_023647.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028150.mm.1 |
NM_023647.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028150.mm.1 |
TC0700003100.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028157.mm.1 |
NM_001256130.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028157.mm.1 |
NM_001256130.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028157.mm.1 |
NM_001256131.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028157.mm.1 |
NM_001256131.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028157.mm.1 |
NM_023647.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028157.mm.1 |
NM_023647.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028157.mm.1 |
TC0700003100.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
NM_001256130.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
NM_001256130.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
NM_001256131.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
NM_001256131.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
NM_001256132.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
NM_001256132.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
NM_001256133.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
NM_001256133.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
NM_023647.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
NM_023647.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028160.mm.1 |
TC0700003100.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700028158.mm.1 |
NM_001256130.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028158.mm.1 |
NM_001256130.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028158.mm.1 |
NM_001256131.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028158.mm.1 |
NM_001256131.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028158.mm.1 |
NM_023647.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028158.mm.1 |
NM_023647.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700028158.mm.1 |
TC0700003100.mm.1 |
MTA TC |