| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600000023.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000023.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000023.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000023.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000023.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000023.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000023.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000027.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000027.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000027.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000027.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000027.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000027.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000027.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000037.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000037.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000037.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000037.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000037.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000037.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000037.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000038.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000038.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000038.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000038.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000038.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000038.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000038.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000042.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000042.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000042.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000042.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000042.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000042.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000042.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000031.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000031.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000031.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000031.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000031.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000031.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000031.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000032.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000032.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000032.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000032.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000032.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000032.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000032.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000026.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000026.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000026.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000026.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000026.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000026.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000026.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000036.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000036.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000036.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000036.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000036.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000036.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000036.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000021.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000021.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000021.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000021.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000021.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000021.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000021.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000025.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000025.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000025.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000025.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000025.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000025.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000025.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000040.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000040.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000040.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000040.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000040.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000040.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000040.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000043.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000043.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000043.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000043.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000043.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000043.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000043.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000020.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000020.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000020.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000020.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000020.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000020.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000020.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000029.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000029.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000029.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000029.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000029.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000029.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000029.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000034.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000034.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000034.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000034.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000034.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000034.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000034.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000030.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000030.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000030.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000030.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000030.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000030.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000030.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000035.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000035.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000035.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000035.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000035.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000035.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000035.mm.1 |
TC0600000004.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600000039.mm.1 |
NM_001167750.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000039.mm.1 |
NM_001167750.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000039.mm.1 |
NM_001167751.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000039.mm.1 |
NM_001167751.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000039.mm.1 |
NM_024260.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000039.mm.1 |
NM_024260.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600000039.mm.1 |
TC0600000004.mm.1 |
MTA TC |