Assay Details
| Gene Symbol: Ccdc132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167750.1 | NP_001161222.1 | 23 - 24 | 2340 | 79 | |
| NM_001167751.1 | NP_001161223.1 | 23 - 24 | 2340 | 79 | ||
| NM_024260.5 | NP_077222.4 | 23 - 24 | 2340 | 79 | ||
| GenBank mRNA | AK042031.1 | - | 23 - 24 | 2320 | 79 | |
| AK075898.1 | - | 23 - 24 | 2329 | 79 | ||
| AK129458.1 | - | 23 - 24 | 2290 | 79 | ||
| AK134533.1 | - | 7 - 8 | 4139 | 79 | ||
| AK217072.1 | - | 1 - 2 | 137 | 79 | ||
| AY254697.1 | - | 23 - 24 | 2333 | 79 | ||
| BC002304.1 | - | 8 - 9 | 885 | 3591600 | 79 | |
| BC036294.1 | - | 23 - 24 | 2280 | 3500316 | 79 | |
| BC043092.1 | - | 23 - 24 | 2297 | 6405356 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc132
Genomic Map