Assay Details
| Gene Symbol: Cyp27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024264.4 | NP_077226.2 | 3 - 4 | 712 | 71 | |
| GenBank mRNA | AK004977.1 | - | 3 - 4 | 713 | 71 | |
| BC002183.1 | - | 2 - 3 | 386 | 3487429 | 71 | |
| BC055028.1 | - | 3 - 4 | 690 | 5098366 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp27a1
Genomic Map