Assay Details
| Gene Symbol: Cyp27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024264.4 | NP_077226.2 | 7 - 8 | 1325 | 90 | |
| GenBank mRNA | AK004977.1 | - | 7 - 8 | 1326 | 90 | |
| BC002183.1 | - | 6 - 7 | 999 | 3487429 | 90 | |
| BC055028.1 | - | 7 - 8 | 1303 | 5098366 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp27a1
Genomic Map