Assay Details
| Gene Symbol: Poc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026173.3 | NP_080449.1 | 4 - 5 | 583 | 76 | |
| GenBank mRNA | AK004769.2 | - | 4 - 5 | 511 | 76 | |
| AK012926.1 | - | 4 - 5 | 551 | 76 | ||
| AK089013.1 | - | 4 - 5 | 519 | 76 | ||
| AK098149.1 | - | 4 - 5 | 476 | 76 | ||
| AK150656.1 | - | 4 - 5 | 584 | 76 | ||
| BC016527.1 | - | 4 - 5 | 497 | 3157281 | 76 | |
| BC027118.1 | - | 4 - 5 | 521 | 5359153 | 76 | |
| BC037631.1 | - | 4 - 5 | 540 | 4954391 | 76 | |
More Information
Set Membership: |
|

Poc5
Genomic Map