Assay Details
| Gene Symbol: Ccdc50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289436.1 | NP_001276365.1 | 8 - 9 | 785 | 114 | |
| NM_026202.4 | NP_080478.2 | 7 - 8 | 739 | 114 | ||
| GenBank mRNA | AJ534985.1 | - | 7 - 8 | 606 | 114 | |
| AK012190.1 | - | 7 - 8 | 633 | 114 | ||
| AK077557.1 | - | 7 - 8 | 677 | 114 | ||
| AK147820.1 | - | 7 - 8 | 690 | 112 | ||
| AK151672.1 | - | 7 - 8 | 701 | 114 | ||
| AK162434.1 | - | 7 - 8 | 716 | 114 | ||
| BC144898.1 | - | 8 - 9 | 765 | 114 | ||
| BC145933.1 | - | 7 - 8 | 718 | 40131236 | 114 | |
| BC145935.1 | - | 7 - 8 | 718 | 40131238 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc50
Genomic Map