Assay Details
| Gene Symbol: 1700019N19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026208.2 | NP_080484.1 | 3 - 4 | 309 | 76 | |
| GenBank mRNA | AK006135.1 | - | 3 - 4 | 310 | 76 | |
| AK006423.1 | - | 3 - 4 | 306 | 76 | ||
| BC049561.1 | - | 3 - 4 | 297 | 6773246 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

1700019N19Rik
Genomic Map