Assay Details
| Gene Symbol: Narfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026238.4 | NP_080514.3 | 1 - 2 | 119 | 100 | |
| GenBank mRNA | AK014038.1 | - | 1 - 2 | 103 | 100 | |
| AK018548.1 | - | 1 - 2 | 90 | 100 | ||
| AK043694.1 | - | 1 - 2 | 96 | 100 | ||
| AK145408.1 | - | 1 - 2 | 122 | 100 | ||
| BC020468.1 | - | 1 - 2 | 79 | 100 | ||
| BC025108.1 | - | 1 - 2 | 99 | 100 | ||
| BC052830.1 | - | 1 - 2 | 96 | 30040958 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Narfl
Genomic Map