Assay Details
| Gene Symbol: Armc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026290.3 | NP_080566.2 | 3 - 4 | 470 | 88 | |
| GenBank mRNA | AK005672.1 | - | 3 - 4 | 467 | 88 | |
| AK015756.1 | - | 3 - 4 | 470 | 88 | ||
| BC049559.1 | - | 3 - 4 | 469 | 6707229 | 88 | |
| EU477370.1 | - | 3 - 4 | 447 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31122919 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31122923 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31122920 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31122925 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302249 | AK015756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302249 | BC049559.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302249 | NM_026290.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302249 | AK005672.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302249 | AK015756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302249 | BC049559.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302249 | NM_026290.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P302249 | AK005672.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 879558 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407421 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 836139 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601393 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168481 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302249 | AK015756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302249 | BC049559.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302249 | NM_026290.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302249 | AK005672.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302249 | AK015756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302249 | BC049559.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302249 | NM_026290.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P302249 | AK005672.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31122953 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168481 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03301049 | - | - |
| Whole Mouse Genome G4122A | A_51_P302249 | AK015756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302249 | BC049559.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302249 | NM_026290.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302249 | AK005672.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302249 | AK015756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302249 | BC049559.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302249 | NM_026290.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P302249 | AK005672.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132210 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118958 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168481 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13549443 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13549435 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13549438 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31122919 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31122920 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31122923 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31122925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31123027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31122988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13385992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13460338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13300652 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13197217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13181445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13231711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13294871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122928 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13549437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13300651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13549435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13385995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13310052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122938 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13385991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13294868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31122956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13528511 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10443370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443377 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443374 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443376 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443375 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443372 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423541_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10443369 | - | - |
| Mouse Genome 430 2.0 Array | 1423541_at | - | - |
| Mouse Genome 430 2.0 Array | 1423541_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67645_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67645_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700004175.mm.1 | NM_026290.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004175.mm.1 | NM_026290.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004175.mm.1 | TC1700000481.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423541_PM_at | - | - |
| Mouse Expression Set 430 | 1423541_at | - | - |
| Murine Genome U74v2 Set | 167439_PM_at | - | - |
| Murine Genome U74v2 Set | 167439_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423541_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423541_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423541_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423541_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 787833 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5900369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005900369 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1212782 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1212782 | - | - |
| mouse-6 v1.1 (Illumina) | scl50920.6.1_80-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5900369 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50920.6.1_80-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5900369 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1212782 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1212782 | - | - |

Armc12
Genomic Map