Assay Details
| Gene Symbol: Mri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026423.4 | NP_080699.3 | 4 - 5 | 1011 | 72 | |
| GenBank mRNA | AK007709.1 | - | 4 - 5 | 972 | 72 | |
| AK010538.1 | - | 4 - 5 | 983 | 72 | ||
| AK013882.1 | - | 4 - 5 | 980 | 72 | ||
| AK047287.1 | - | 4 - 5 | 1014 | 72 | ||
| AK144807.1 | - | 4 - 5 | 954 | 72 | ||
| AK164894.1 | - | 4 - 5 | 980 | 72 | ||
| BC025049.1 | - | 4 - 5 | 978 | 4951045 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular amino acid biosynthetic process
methionine biosynthetic process
L-methionine biosynthetic process from methylthioadenosine
cellular metabolic process
cellular biosynthetic process
isomerase activity
identical protein binding
S-methyl-5-thioribose-1-phosphate isomerase activity
cellular amino acid biosynthetic process
methionine biosynthetic process
L-methionine biosynthetic process from methylthioadenosine
cellular metabolic process
cellular biosynthetic process
isomerase activity
identical protein binding
S-methyl-5-thioribose-1-phosphate isomerase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26019018 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26019014 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26019015 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26019017 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26019016 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P367791 | AK164894.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P367791 | BC009164.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P367791 | BC025049.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P367791 | AK013882.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P367791 | AK047287.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P367791 | AK164894.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P367791 | BC009164.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P367791 | BC025049.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P367791 | AK013882.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P367791 | AK047287.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784788 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411184 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117388 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P367791 | AK164894.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P367791 | BC009164.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P367791 | BC025049.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P367791 | AK013882.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P367791 | AK047287.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P367791 | AK164894.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P367791 | BC009164.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P367791 | BC025049.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P367791 | AK013882.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P367791 | AK047287.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26019019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26018987 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106181 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01783131 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06577722 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117388 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01783137 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06577719 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01783221 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112621 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01783108 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06577817 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01783203 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01783096 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01783126 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01783122 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01783115 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01783125 | - | - |
| Whole Mouse Genome G4122A | A_52_P535782 | AK047287.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367792 | AK164894.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367792 | BC025049.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367792 | AK013882.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367792 | AK047287.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367792 | AK144807.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367791 | AK164894.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367791 | BC009164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367791 | BC025049.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367791 | AK013882.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367791 | AK047287.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367791 | AK164894.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367791 | BC009164.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367791 | BC025049.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367791 | AK013882.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367791 | AK047287.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103104 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136647 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06577719 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117388 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01783131 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106181 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01783115 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112621 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07308039 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07269032 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07429725 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07010999 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07308043 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07429724 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26019015 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26019017 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26019014 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26019018 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26019016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26019018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26018987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06979530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06855856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07436040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07436035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26018996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26019019 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10580164 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580163 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580162 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580161 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580166 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580165 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10580168 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426549_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417146_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10580160 | - | - |
| Mouse Genome 430 2.0 Array | 1417146_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417146_at | - | - |
| Mouse Genome 430 2.0 Array | 1426549_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426549_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67873_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67873_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800019660.mm.1 | NM_026423.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019660.mm.1 | NM_026423.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019660.mm.1 | TC0800002579.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800019661.mm.1 | NM_026423.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019661.mm.1 | NM_026423.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019661.mm.1 | TC0800002579.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800019659.mm.1 | NM_026423.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019659.mm.1 | NM_026423.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019659.mm.1 | TC0800002579.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800019658.mm.1 | NM_026423.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019658.mm.1 | NM_026423.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019658.mm.1 | TC0800002579.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800019662.mm.1 | NM_026423.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019662.mm.1 | NM_026423.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800019662.mm.1 | TC0800002579.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417146_PM_at | - | - |
| Mouse Expression Set 430 | 1417146_at | - | - |
| Mouse Expression Set 430 | 1426549_PM_at | - | - |
| Mouse Expression Set 430 | 1426549_at | - | - |
| Murine Genome U74v2 Set | 169759_r_at | - | - |
| Murine Genome U74v2 Set | 169759_PM_r_at | - | - |
| Murine Genome U74v2 Set | 165505_at | - | - |
| Murine Genome U74v2 Set | 165505_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426549_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417146_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426549_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417146_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426549_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417146_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426549_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417146_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 660935 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5910168 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005910168 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2684360 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2684360 | - | - |
| mouse-6 v1.1 (Illumina) | scl34566.5_165-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5910168 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34566.5_165-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5910168 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2844848 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2844848 | - | - |

Mri1
Genomic Map