| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200007299.mm.1 |
NM_001177841.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007299.mm.1 |
NM_001177841.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007299.mm.1 |
TC1200001028.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200007292.mm.1 |
NR_033587.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007292.mm.1 |
NR_033587.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007292.mm.1 |
TC1200001028.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200007298.mm.1 |
NM_001177841.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007298.mm.1 |
NM_001177841.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007298.mm.1 |
TC1200001028.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200007303.mm.1 |
NM_001177841.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007303.mm.1 |
NM_001177841.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007303.mm.1 |
NM_026580.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007303.mm.1 |
NM_026580.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007303.mm.1 |
NR_033587.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007303.mm.1 |
NR_033587.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200007303.mm.1 |
TC1200001028.mm.1 |
MTA TC |