Assay Details
| Gene Symbol: Tescl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163810.1 | NP_001157282.1 | 1 | 76 | 72 | |
| GenBank mRNA | AK005787.1 | - | 1 | 50 | 72 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK005787.1,NM_001163810.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010216 | NM_001163810.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010216 | AK005787.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010216 | BC125456.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010216 | BC125458.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605572 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010216 | NM_001163810.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010216 | AK005787.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010216 | BC125456.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010216 | BC125458.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | NM_001163810.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P128075 | AK005787.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06619807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06619799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06064282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05915394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06009996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06221517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06619812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06009993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06619795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06619808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06406541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05885227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05915391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004372 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1449284_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560851 | - | - |
| Mouse Genome 430 2.0 Array | 1449284_at | - | - |
| Mouse Genome 430 2.0 Array | 1449284_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69301_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69301_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700022445.mm.1 | NM_001163810.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022445.mm.1 | NM_001163810.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022445.mm.1 | TC0700002597.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449284_PM_at | - | - |
| Mouse Expression Set 430 | 1449284_at | - | - |
| Murine Genome U74v2 Set | 165902_PM_at | - | - |
| Murine Genome U74v2 Set | 165902_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449284_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449284_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449284_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449284_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 776962 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5550017 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2631903 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2631903 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2631903 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631903 | - | - |

Tescl
Genomic Map