Assay Details
| Gene Symbol: Mus81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027877.3 | NP_082153.3 | 4 - 5 | 659 | 74 | |
| GenBank mRNA | AF425647.1 | - | 4 - 5 | 474 | 74 | |
| AK004647.1 | - | 3 - 4 | 342 | 74 | ||
| AK145858.1 | - | 4 - 5 | 609 | 74 | ||
| AK205475.1 | - | 2 - 3 | 122 | 74 | ||
| BC026560.1 | - | 4 - 5 | 672 | 5038349 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
DNA catabolic process, endonucleolytic
DNA metabolic process
DNA repair
DNA recombination
cellular response to DNA damage stimulus
response to intra-S DNA damage checkpoint signaling
DNA binding
nuclease activity
endonuclease activity
hydrolase activity
metal ion binding
3'-flap endonuclease activity
DNA catabolic process, endonucleolytic
DNA metabolic process
DNA repair
DNA recombination
cellular response to DNA damage stimulus
response to intra-S DNA damage checkpoint signaling
DNA binding
nuclease activity
endonuclease activity
hydrolase activity
metal ion binding
3'-flap endonuclease activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31929033 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31929031 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406507 | BC026560.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406507 | AK145858.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406507 | NM_027877.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406507 | AK004647.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406507 | BC026560.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406507 | AK145858.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406507 | NM_027877.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P406507 | AK004647.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128608 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161245 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406507 | BC026560.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406507 | AK145858.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406507 | NM_027877.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406507 | AK004647.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406507 | BC026560.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406507 | AK145858.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406507 | NM_027877.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P406507 | AK004647.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P114372 | AF425647.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P406507 | BC026560.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P406507 | AK145858.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P406507 | NM_027877.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P406507 | AK004647.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P406507 | BC026560.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P406507 | AK145858.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P406507 | NM_027877.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P406507 | AK004647.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31929018 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161245 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09093574 | - | - |
| Whole Mouse Genome G4122A | A_51_P114372 | AF425647.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P406507 | BC026560.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P406507 | AK145858.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P406507 | NM_027877.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P406507 | AK004647.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P406507 | BC026560.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P406507 | AK145858.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P406507 | NM_027877.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P406507 | AK004647.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119743 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109512 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161245 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14230222 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14230217 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14230226 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14493578 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14230210 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14230228 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14230219 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14230211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31929031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31929033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31929005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14230228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14511270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14230210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14230218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14234265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14511274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14234267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31929020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928972 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465075 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465079 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465076 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465083 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465073 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465078 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465077 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465074 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465080 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450432_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431885_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465072 | - | - |
| Mouse Genome 430 2.0 Array | 1431885_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450432_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1431885_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450432_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71711_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71711_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900008645.mm.1 | NM_027877.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900008645.mm.1 | NM_027877.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900008645.mm.1 | TC1900000969.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431885_PM_a_at | - | - |
| Mouse Expression Set 430 | 1431885_a_at | - | - |
| Mouse Expression Set 430 | 1450432_s_at | - | - |
| Mouse Expression Set 430 | 1450432_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450432_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431885_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450432_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431885_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450432_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450432_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431885_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431885_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 443285 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6660184 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660184 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2651462 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2651462 | - | - |
| mouse-6 v1.1 (Illumina) | scl53497.9_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660184 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53497.9_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660184 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2651462 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2957665 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2651462 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2957665 | - | - |

Mus81
Genomic Map