Assay Details
Gene Symbol: C1qtnf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204134.1 | NP_001191063.1 | 4 - 5 | 796 | 85 | |
NM_030888.4 | NP_112150.1 | 4 - 5 | 602 | 85 | ||
GenBank mRNA | AF246265.1 | - | 4 - 5 | 579 | 85 | |
AK029352.1 | - | 4 - 5 | 579 | 85 | ||
AK075867.1 | - | 4 - 5 | 579 | 85 | ||
AK081704.1 | - | 4 - 5 | 579 | 85 | ||
AK128898.1 | - | 4 - 5 | 602 | 85 | ||
BC019636.1 | - | 4 - 5 | 805 | 85 | ||
BC055047.1 | - | 4 - 5 | 564 | 6486507 | 85 | |
BC055859.1 | - | 4 - 5 | 543 | 4238617 | 85 | |
DQ002396.1 | - | 4 - 5 | 480 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
