Assay Details
| Gene Symbol: Smarcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130187.1 | NP_001123659.1 | 4 - 5 | 832 | 101 | |
| NM_031878.2 | NP_114084.2 | 3 - 4 | 466 | 101 | ||
| GenBank mRNA | AK159259.1 | - | 4 - 5 | 669 | 101 | |
| BC005732.1 | - | 3 - 4 | 456 | 2647494 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smarcd2
Genomic Map