Assay Details
| Gene Symbol: Npnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287101.1 | NP_001274030.1 | 2 - 3 | 421 | 96 | |
| NM_033525.3 | NP_277060.2 | 2 - 3 | 421 | 96 | ||
| GenBank mRNA | AF397008.1 | - | 2 - 3 | 173 | 96 | |
| AI527763.1 | - | 2 - 3 | 301 | 96 | ||
| AY035899.1 | - | 2 - 3 | 296 | 96 | ||
| CK387038.1 | - | 2 - 3 | 295 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npnt
Genomic Map