Assay Details
| Gene Symbol: Cd163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170395.1 | NP_001163866.1 | 6 - 7 | 1509 | 83 | |
| NM_053094.2 | NP_444324.2 | 6 - 7 | 1509 | 83 | ||
| GenBank mRNA | AF274883.1 | - | 6 - 7 | 1504 | 83 | |
| BC144848.1 | - | 6 - 7 | 1460 | 9053396 | 83 | |
| BC145793.1 | - | 6 - 7 | 1460 | 40131096 | 83 | |
| DQ058616.1 | - | 6 - 7 | 1465 | 83 | ||
| DQ058617.1 | - | 6 - 7 | 1465 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd163
Genomic Map