Assay Details
| Gene Symbol: Acss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080575.2 | NP_542142.1 | 6 - 7 | 1130 | 66 | |
| GenBank mRNA | AB046742.1 | - | 6 - 7 | 1119 | 66 | |
| AK088244.1 | - | 6 - 7 | 1109 | 66 | ||
| AK146568.1 | - | 6 - 7 | 1131 | 66 | ||
| AK158119.1 | - | 6 - 7 | 1125 | 66 | ||
| AK172412.1 | - | 6 - 7 | 1116 | 66 | ||
| AK172470.1 | - | 6 - 7 | 1105 | 66 | ||
| AK173275.1 | - | 6 - 7 | 1091 | 66 | ||
| BC030930.1 | - | 2 - 3 | 304 | 4911679 | 66 | |
| BC092278.1 | - | 6 - 7 | 1159 | 3710505 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acss1
Genomic Map