Assay Details
| Gene Symbol: Pecam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037551.1 | - | 10 - 11 | 2473 | 61 | |
| AK084527.1 | - | 9 - 10 | 2063 | 61 | ||
| AK169431.1 | - | 8 - 9 | 1762 | 61 | ||
| BC008519.1 | - | 9 - 10 | 2067 | 3592726 | 61 | |
| BC085502.1 | - | 9 - 10 | 1869 | 5720681 | 61 | |
| L06039.1 | - | 9 - 10 | 1952 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pecam1
Genomic Map