Assay Details
| Gene Symbol: Rhox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008818.2 | NP_032844.2 | 118 | 67 | ||
| GenBank mRNA | AK146379.1 | - | 143 | 67 | ||
| AK146393.1 | - | 142 | 67 | |||
| AK146449.1 | - | 124 | 67 | |||
| BC056204.1 | - | 2 | 126 | 30022528 | 67 | |
| DQ058642.1 | - | 102 | 67 | |||
| M32484.1 | - | 102 | 67 | |||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK146379.1,AK146393.1,AK146449.1,BC056204.1,DQ058642.1,M32484.1,NM_008818.2 transcripts.
Set Membership: |
|

Rhox5
Genomic Map