Assay Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053943.1 | - | 13 - 14 | 1806 | 62 | |
| AK004712.1 | - | 9 - 10 | 815 | 62 | ||
| AK128980.1 | - | 13 - 14 | 1882 | 62 | ||
| AK158276.1 | - | 13 - 14 | 1833 | 62 | ||
| AK159330.1 | - | 13 - 14 | 1813 | 62 | ||
| AK159342.1 | - | 13 - 14 | 1832 | 62 | ||
| AK159377.1 | - | 13 - 14 | 1803 | 62 | ||
| AK159409.1 | - | 13 - 14 | 1833 | 62 | ||
| AK159774.1 | - | 13 - 14 | 1804 | 62 | ||
| AK159957.1 | - | 13 - 14 | 1804 | 62 | ||
| AK187128.1 | - | 1 - 2 | 71 | 62 | ||
| BC082577.1 | - | 13 - 14 | 1818 | 30648530 | 62 | |
| X80478.1 | - | 4 - 5 | 536 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aebp1
Genomic Map