Assay Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053943.1 | - | 14 - 15 | 1887 | 80 | |
| AK004712.1 | - | 10 - 11 | 896 | 80 | ||
| AK128980.1 | - | 14 - 15 | 1963 | 80 | ||
| AK158276.1 | - | 14 - 15 | 1914 | 80 | ||
| AK159330.1 | - | 14 - 15 | 1894 | 80 | ||
| AK159342.1 | - | 14 - 15 | 1913 | 80 | ||
| AK159377.1 | - | 14 - 15 | 1884 | 80 | ||
| AK159409.1 | - | 14 - 15 | 1914 | 80 | ||
| AK159774.1 | - | 14 - 15 | 1885 | 80 | ||
| AK159957.1 | - | 14 - 15 | 1885 | 80 | ||
| AK187128.1 | - | 2 - 3 | 152 | 80 | ||
| AK193273.1 | - | 1 - 2 | 26 | 80 | ||
| BC082577.1 | - | 14 - 15 | 1899 | 30648530 | 80 | |
| X80478.1 | - | 5 - 6 | 617 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aebp1
Genomic Map