Assay Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053943.1 | - | 17 - 18 | 2392 | 67 | |
| AK004712.1 | - | 13 - 14 | 1401 | 67 | ||
| AK128980.1 | - | 17 - 18 | 2468 | 67 | ||
| AK158276.1 | - | 17 - 18 | 2419 | 67 | ||
| AK159330.1 | - | 17 - 18 | 2399 | 67 | ||
| AK159342.1 | - | 17 - 18 | 2418 | 67 | ||
| AK159377.1 | - | 17 - 18 | 2389 | 67 | ||
| AK159409.1 | - | 17 - 18 | 2419 | 67 | ||
| AK159774.1 | - | 17 - 18 | 2390 | 67 | ||
| AK159957.1 | - | 17 - 18 | 2390 | 67 | ||
| BC082577.1 | - | 17 - 18 | 2404 | 30648530 | 67 | |
| X80478.1 | - | 8 - 9 | 1122 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aebp1
Genomic Map