Assay Details
| Gene Symbol: Btrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037758.2 | NP_001032847.2 | 5 - 6 | 660 | 71 | |
| NM_001286465.1 | NP_001273394.1 | 5 - 6 | 660 | 71 | ||
| NM_009771.3 | NP_033901.1 | 4 - 5 | 552 | 71 | ||
| GenBank mRNA | AF081887.1 | - | 4 - 5 | 537 | 71 | |
| AF099932.1 | - | 4 - 5 | 447 | 71 | ||
| AF112979.1 | - | 4 - 5 | 447 | 71 | ||
| AK032221.1 | - | 5 - 6 | 623 | 71 | ||
| AK052317.1 | - | 6 - 7 | 684 | 71 | ||
| AK083257.1 | - | 5 - 6 | 636 | 71 | ||
| AK145624.1 | - | 5 - 6 | 643 | 71 | ||
| AK156660.1 | - | 4 - 5 | 580 | 71 | ||
| AK220183.1 | - | 5 - 6 | 658 | 71 | ||
| BC003989.1 | - | 4 - 5 | 453 | 3491843 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Btrc
Genomic Map