Assay Details
| Gene Symbol: Prl7a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011168.4 | NP_035298.1 | 4 - 5 | 403 | 67 | |
| GenBank mRNA | AF011382.1 | - | 4 - 5 | 392 | 67 | |
| AF020524.1 | - | 4 - 5 | 399 | 67 | ||
| AK005455.1 | - | 4 - 5 | 404 | 67 | ||
| AK028068.1 | - | 4 - 5 | 403 | 67 | ||
| AK131665.1 | - | 4 - 5 | 404 | 67 | ||
| BC099468.1 | - | 4 - 5 | 382 | 30789902 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102798 | AK131665.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2102798 | NM_011168.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555185 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 607417 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602870 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925330 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741992 | AK032645.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741992 | AK034716.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741992 | AK035452.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741992 | AK128894.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741992 | BC006717.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741992 | BC082602.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 741992 | NM_001114328.1 | refseq_mrna_mouse_NM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163651 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102798 | AK131665.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2102798 | NM_011168.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28903122 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163651 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07819024 | - | - |
| Whole Mouse Genome G4122A | A_51_P236074 | AF011382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P236074 | AK005455.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P236074 | AK028068.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P236074 | AK131665.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P236074 | BC099468.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116604 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07819024 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163651 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11058419 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11012682 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11012680 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11012683 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11012685 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11012687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28903170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10455608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10916556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11092157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11115835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10437598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11058419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10845711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28903155 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408435 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10408434 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417905_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408429 | - | - |
| Mouse Genome 430 2.0 Array | 1417905_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417905_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19114_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19114_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300010560.mm.1 | NM_011168.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010560.mm.1 | NM_011168.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300010560.mm.1 | TC1300001696.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417905_PM_at | - | - |
| Mouse Expression Set 430 | 1417905_at | - | - |
| Murine Genome U74v2 Set | 97946_at | - | - |
| Murine Genome U74v2 Set | 97946_PM_at | - | - |
| Murine Genome U74v2 Set | 162303_PM_f_at | - | - |
| Murine Genome U74v2 Set | 162303_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417905_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417905_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417905_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417905_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1940402 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001940402 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220021 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220021 | - | - |
| mouse-6 v1.1 (Illumina) | scl44166.6.1_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1940402 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44166.6.1_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940402 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1220021 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220021 | - | - |

Prl7a2
Genomic Map