Assay Details
| Gene Symbol: Pts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011220.2 | NP_035350.1 | 1 - 2 | 84 | 95 | |
| GenBank mRNA | AF043225.1 | - | 1 - 2 | 99 | 95 | |
| AK002614.1 | - | 1 - 2 | 94 | 95 | ||
| AK146987.1 | - | 1 - 2 | 95 | 95 | ||
| AK153959.1 | - | 1 - 2 | 100 | 95 | ||
| BC029013.1 | - | 1 - 2 | 84 | 5357873 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504100 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504102 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504098 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177021 | BC029013.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177021 | AK153959.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177021 | AK002614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177021 | AK146987.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177021 | NM_011220.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177021 | BC029013.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177021 | AK153959.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177021 | AK002614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177021 | AK146987.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P177021 | NM_011220.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237712 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 170862 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100191 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177021 | BC029013.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177021 | AK153959.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177021 | AK002614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177021 | AK146987.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177021 | NM_011220.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177021 | BC029013.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177021 | AK153959.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177021 | AK002614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177021 | AK146987.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P177021 | NM_011220.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204295 | BC029013.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204295 | AK153959.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204295 | AK002614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204295 | AK146987.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P204295 | NM_011220.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P177021 | BC029013.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P177021 | AK153959.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P177021 | AK002614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P177021 | AK146987.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P177021 | NM_011220.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P177021 | BC029013.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P177021 | AK153959.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P177021 | AK002614.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P177021 | AK146987.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P177021 | NM_011220.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504101 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26504103 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100191 | - | - |
| Whole Mouse Genome G4122A | A_52_P177021 | BC029013.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177021 | AK153959.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177021 | AK002614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177021 | AK146987.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177021 | NM_011220.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177021 | BC029013.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177021 | AK153959.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177021 | AK002614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177021 | AK146987.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P177021 | NM_011220.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204295 | BC029013.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204295 | AK153959.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204295 | AK002614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204295 | AK146987.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P204295 | NM_011220.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129313 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100191 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07867587 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08168657 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08168656 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08168650 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08168658 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504099 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504102 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504101 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504098 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26504100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26504059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07998467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504061 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07654655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07654654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07744282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08168650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08168656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08168658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07583419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26504123 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10593329 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593327 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593328 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593330 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593331 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593326 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458402_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450660_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10593325 | - | - |
| Mouse Genome 430 2.0 Array | 1450660_at | - | - |
| Mouse Genome 430 2.0 Array | 1458402_at | - | - |
| Mouse Genome 430 2.0 Array | 1450660_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458402_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19286_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19286_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900018855.mm.1 | NM_011220.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018855.mm.1 | NM_011220.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018855.mm.1 | TC0900002268.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900018856.mm.1 | NM_011220.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018856.mm.1 | NM_011220.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900018856.mm.1 | TC0900002268.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450660_PM_at | - | - |
| Mouse Expression Set 430 | 1450660_at | - | - |
| Mouse Expression Set 430 | 1458402_PM_at | - | - |
| Mouse Expression Set 430 | 1458402_at | - | - |
| Murine Genome U74v2 Set | 160844_PM_at | - | - |
| Murine Genome U74v2 Set | 160844_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450660_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450660_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450660_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450660_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458402_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458402_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 910187 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1500270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1500270 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001500270 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217378 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1217378 | - | - |
| mouse-6 v1.1 (Illumina) | scl35880.1.84_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1500270 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl35880.1.84_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1500270 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1217378 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2848548 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217378 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2848548 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl35880.1.84_1-S | - | - |

Pts
Genomic Map