Assay Details
| Gene Symbol: Naglu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013792.2 | NP_038820.2 | 1 - 2 | 407 | 56 | |
| GenBank mRNA | AK143546.1 | - | 1 - 2 | 408 | 56 | |
| AK164125.1 | - | 1 - 2 | 386 | 56 | ||
| AK170758.1 | - | 1 - 2 | 390 | 56 | ||
| BC055733.1 | - | 1 - 2 | 381 | 6411530 | 56 | |
| U85247.1 | - | 1 - 2 | 376 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Naglu
Genomic Map