Assay Details
| Gene Symbol: Naglu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013792.2 | NP_038820.2 | 3 - 4 | 708 | 73 | |
| GenBank mRNA | AK143546.1 | - | 3 - 4 | 709 | 73 | |
| AK164125.1 | - | 3 - 4 | 687 | 73 | ||
| AK170758.1 | - | 3 - 4 | 691 | 73 | ||
| BC055733.1 | - | 3 - 4 | 682 | 6411530 | 73 | |
| U85247.1 | - | 3 - 4 | 677 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Naglu
Genomic Map