| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800011982.mm.1 |
NM_001110100.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011982.mm.1 |
NM_001285981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011982.mm.1 |
NM_001285983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011982.mm.1 |
NM_016812.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011982.mm.1 |
TC0800001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800011994.mm.1 |
NM_001110100.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011994.mm.1 |
NM_001285981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011994.mm.1 |
NM_001285983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011994.mm.1 |
NM_016812.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011994.mm.1 |
TC0800001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800011996.mm.1 |
NM_001110100.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011996.mm.1 |
NM_001285981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011996.mm.1 |
NM_001285983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011996.mm.1 |
NM_016812.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011996.mm.1 |
TC0800001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800011985.mm.1 |
NM_001110100.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011985.mm.1 |
NM_001285981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011985.mm.1 |
NM_001285983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011985.mm.1 |
NM_016812.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011985.mm.1 |
TC0800001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800011995.mm.1 |
NM_001110100.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011995.mm.1 |
NM_001285981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011995.mm.1 |
NM_001285983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011995.mm.1 |
NM_016812.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011995.mm.1 |
TC0800001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800011990.mm.1 |
NM_001110100.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011990.mm.1 |
NM_001285981.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011990.mm.1 |
NM_001285983.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011990.mm.1 |
NM_016812.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800011990.mm.1 |
TC0800001477.mm.1 |
MTA TC |