Assay Details
| Gene Symbol: Acss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019811.3 | NP_062785.2 | 10 - 11 | 1347 | 90 | |
| GenBank mRNA | AF216873.1 | - | 10 - 11 | 1297 | 90 | |
| AK028586.1 | - | 10 - 11 | 1325 | 90 | ||
| AK029014.1 | - | 10 - 11 | 1315 | 90 | ||
| AK029262.1 | - | 1333 | 90 | |||
| AK053877.1 | - | 10 - 11 | 1321 | 90 | ||
| AK093831.1 | - | 10 - 11 | 1324 | 90 | ||
| BC040790.1 | - | 10 - 11 | 1297 | 90 | ||
| BC051432.1 | - | 10 - 11 | 1347 | 6515568 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acss2
Genomic Map