Assay Details
| Gene Symbol: Pcdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021543.4 | NP_067518.2 | 1 - 2 | 2819 | 119 | |
| GenBank mRNA | AF231125.1 | - | 1 - 2 | 2743 | 119 | |
| BC053008.1 | - | 1 - 2 | 2807 | 5698009 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcdh8
Genomic Map