Assay Details
| Gene Symbol: Tmub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022418.3 | NP_071863.1 | 2 - 3 | 510 | 70 | |
| GenBank mRNA | AB030183.1 | - | 2 - 3 | 472 | 70 | |
| AK008109.1 | - | 2 - 3 | 444 | 70 | ||
| AY603379.1 | - | 2 - 3 | 441 | 70 | ||
| BC019547.1 | - | 2 - 3 | 466 | 4164765 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541914 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541913 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957815 | BC019547.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957815 | NM_022418.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957815 | AB030183.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957815 | AW125847.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732174 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300180 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179526 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957815 | BC019547.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957815 | NM_022418.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957815 | AB030183.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957815 | AW125847.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23541905 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179526 | - | - |
| Whole Mouse Genome G4122A | A_51_P385390 | BC019547.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385390 | NM_022418.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385390 | AB030183.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385390 | AK008109.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P385390 | AW125847.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111406 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179526 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04214868 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04161714 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04432611 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04432619 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04161717 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04948082 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04214860 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04432625 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04432613 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04412874 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541914 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23541913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23541862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04161716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04617760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04804530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04432626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23541912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04432613 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10528613 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528614 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528612 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528611 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448840_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528610 | - | - |
| Mouse Genome 430 2.0 Array | 1448840_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448840_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64295_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64295_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500018941.mm.1 | NM_022418.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018941.mm.1 | NM_022418.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018941.mm.1 | TC0500002079.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500018942.mm.1 | NM_022418.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018942.mm.1 | NM_022418.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500018942.mm.1 | TC0500002079.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448840_at | - | - |
| Mouse Expression Set 430 | 1448840_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448840_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448840_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448840_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448840_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 385382 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4610068 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610068 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2730829 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2730829 | - | - |
| mouse-6 v1.1 (Illumina) | scl26803.4.1_96-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610068 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26803.4.1_96-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610068 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2730829 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2932660 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2932660 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2730829 | - | - |

Tmub1
Genomic Map