Assay Details
| Gene Symbol: Cwc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023153.3 | NP_075642.1 | 3 - 4 | 411 | 98 | |
| GenBank mRNA | AJ250394.1 | - | 3 - 4 | 302 | 98 | |
| AK002864.1 | - | 3 - 4 | 350 | 98 | ||
| AK003202.1 | - | 3 - 4 | 343 | 98 | ||
| AK003591.1 | - | 1 - 2 | 53 | 98 | ||
| AK003834.2 | - | 3 - 4 | 356 | 98 | ||
| AK145249.1 | - | 3 - 4 | 351 | 98 | ||
| AK167776.1 | - | 3 - 4 | 331 | 98 | ||
| AK168365.1 | - | 3 - 4 | 331 | 98 | ||
| AY842452.1 | - | 2 - 3 | 257 | 98 | ||
| BC004726.1 | - | 3 - 4 | 347 | 3582653 | 98 | |
| BQ840384.1 | - | 3 - 4 | 411 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26317494 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26317495 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445307 | AK167776.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445307 | AK168365.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445307 | AY842452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445307 | BC004726.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445307 | AK002864.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445307 | AK167776.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445307 | AK168365.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445307 | AY842452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445307 | BC004726.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P445307 | AK002864.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2139894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587608 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 809687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73126 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104982 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445307 | AK167776.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445307 | AK168365.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445307 | AY842452.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445307 | BC004726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445307 | AK002864.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445307 | AK167776.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445307 | AK168365.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445307 | AY842452.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445307 | BC004726.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P445307 | AK002864.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2139894 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508947 | AK167776.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508947 | AK168365.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508947 | AY842452.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508947 | BC004726.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508947 | AK002864.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26317499 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104982 | - | - |
| Whole Mouse Genome G4122A | A_51_P508947 | AK167776.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508947 | AK168365.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508947 | AY842452.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508947 | BC004726.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508947 | AK002864.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445307 | AK167776.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445307 | AK168365.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445307 | AY842452.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445307 | BC004726.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445307 | AK002864.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445307 | AK167776.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445307 | AK168365.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445307 | AY842452.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445307 | BC004726.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445307 | AK002864.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126569 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110553 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104982 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07956999 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07566177 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07957000 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26317495 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26317494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26317560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08152091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07548596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07662547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07674809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08177381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08049593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07674811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07957000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08049594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07999083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07548604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26317512 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10583260 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583258 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583261 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583259 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10583256 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423133_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10583254 | - | - |
| Mouse Genome 430 2.0 Array | 1423133_at | - | - |
| Mouse Genome 430 2.0 Array | 1423133_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66070_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66070_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900000749.mm.1 | NM_023153.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000749.mm.1 | NM_023153.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000749.mm.1 | TC0900000105.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900000748.mm.1 | NM_023153.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000748.mm.1 | NM_023153.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900000748.mm.1 | TC0900000105.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423133_at | - | - |
| Mouse Expression Set 430 | 1423133_PM_at | - | - |
| Murine Genome U74v2 Set | 99615_at | - | - |
| Murine Genome U74v2 Set | 99616_PM_s_at | - | - |
| Murine Genome U74v2 Set | 99615_PM_at | - | - |
| Murine Genome U74v2 Set | 99616_s_at | - | - |
| Murine Genome U74v2 Set | 170200_i_at | - | - |
| Murine Genome U74v2 Set | 170200_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423133_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423133_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423133_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423133_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 917783 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5360672 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5360672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005360672 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2663410 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2663410 | - | - |
| mouse-6 v1.1 (Illumina) | scl066070.9_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5360672 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066070.9_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5360672 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2663410 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2663410 | - | - |

Cwc15
Genomic Map