| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400015593.mm.1 |
NM_001085473.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015593.mm.1 |
NM_001085473.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015593.mm.1 |
NM_023190.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015593.mm.1 |
NM_023190.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015593.mm.1 |
TC1400002063.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400015588.mm.1 |
NM_001085473.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015588.mm.1 |
NM_001085473.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015588.mm.1 |
NM_023190.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015588.mm.1 |
NM_023190.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015588.mm.1 |
TC1400002063.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400015590.mm.1 |
NM_001085473.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015590.mm.1 |
NM_001085473.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015590.mm.1 |
NM_023190.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015590.mm.1 |
NM_023190.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015590.mm.1 |
TC1400002063.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400015596.mm.1 |
NM_001085473.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015596.mm.1 |
NM_001085473.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015596.mm.1 |
NM_023190.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015596.mm.1 |
NM_023190.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015596.mm.1 |
TC1400002063.mm.1 |
MTA TC |