Assay Details
| Gene Symbol: Pcbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025273.4 | NP_079549.1 | 3 - 4 | 299 | 72 | |
| GenBank mRNA | AK007401.1 | - | 3 - 4 | 271 | 72 | |
| BC024354.1 | - | 3 - 4 | 232 | 5052788 | 72 | |
| BY116433.1 | - | 2 - 3 | 167 | 72 | ||
| BY294059.1 | - | 3 - 4 | 287 | 72 | ||
| M83741.1 | - | 2 - 3 | 209 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
tetrahydrobiopterin biosynthetic process
regulation of protein homodimerization activity
positive regulation of transcription, DNA-templated
protein homotetramerization
protein heterooligomerization
oxidation-reduction process
transcription coactivator activity
phenylalanine 4-monooxygenase activity
protein binding
4-alpha-hydroxytetrahydrobiopterin dehydratase activity
lyase activity
identical protein binding
transcription, DNA-templated
regulation of transcription, DNA-templated
tetrahydrobiopterin biosynthetic process
regulation of protein homodimerization activity
positive regulation of transcription, DNA-templated
protein homotetramerization
protein heterooligomerization
oxidation-reduction process
transcription coactivator activity
phenylalanine 4-monooxygenase activity
protein binding
4-alpha-hydroxytetrahydrobiopterin dehydratase activity
lyase activity
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27178540 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27178539 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27178541 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139320 | M83741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139320 | NM_025273.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139320 | AK007401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139320 | BC024354.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139320 | CF355072.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139320 | M83741.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139320 | NM_025273.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139320 | AK007401.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139320 | BC024354.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P139320 | CF355072.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 803298 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910174 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139331 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139320 | M83741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139320 | NM_025273.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139320 | AK007401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139320 | BC024354.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139320 | CF355072.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139320 | M83741.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139320 | NM_025273.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139320 | AK007401.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139320 | BC024354.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P139320 | CF355072.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139320 | M83741.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139320 | NM_025273.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139320 | AK007401.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139320 | BC024354.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139320 | CF355072.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139320 | M83741.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139320 | NM_025273.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139320 | AK007401.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139320 | BC024354.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P139320 | CF355072.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27178554 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139331 | - | - |
| Whole Mouse Genome G4122A | A_51_P139320 | M83741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139320 | NM_025273.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139320 | AK007401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139320 | BC024354.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139320 | CF355072.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139320 | M83741.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139320 | NM_025273.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139320 | AK007401.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139320 | BC024354.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P139320 | CF355072.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103988 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139331 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08351344 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08351338 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08670653 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08351333 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08670654 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08351345 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27178539 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27178540 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27178541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27178559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08931465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08761889 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08433051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178567 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08917097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08760667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08867709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08516672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08917099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08670653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08670654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08296246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08752952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08351345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27178561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08413871 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10363457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10363459 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10363458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10363456 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418713_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10363455 | - | - |
| Mouse Genome 430 2.0 Array | 1418713_at | - | - |
| Mouse Genome 430 2.0 Array | 1418713_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13180_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 13180_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000003788.mm.1 | NM_025273.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000003788.mm.1 | TC1000000558.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418713_at | - | - |
| Mouse Expression Set 430 | 1418713_PM_at | - | - |
| Murine Genome U74v2 Set | 99056_PM_at | - | - |
| Murine Genome U74v2 Set | 99056_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418713_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418713_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418713_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418713_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 601254 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 130148 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 130148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000130148 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2690575 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2690575 | - | - |
| mouse-6 v1.1 (Illumina) | scl38875.4.1_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 130148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38875.4.1_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 130148 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2690575 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2690575 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl38875.4.1_6-S | - | - |

Pcbd1
Genomic Map