Assay Details
| Gene Symbol: Tmem14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025387.3 | NP_079663.1 | 5 - 6 | 472 | 67 | |
| GenBank mRNA | AK003891.1 | - | 5 - 6 | 421 | 67 | |
| AK008495.1 | - | 4 - 5 | 362 | 67 | ||
| AK008878.1 | - | 5 - 6 | 394 | 67 | ||
| AK010609.1 | - | 5 - 6 | 432 | 67 | ||
| AK012101.1 | - | 5 - 6 | 419 | 67 | ||
| AK019395.1 | - | 5 - 6 | 421 | 67 | ||
| AK050688.1 | - | 5 - 6 | 407 | 67 | ||
| AK089021.1 | - | 4 - 5 | 361 | 67 | ||
| AK217054.1 | - | 1 - 2 | 66 | 67 | ||
| BC025854.1 | - | 5 - 6 | 373 | 5250659 | 67 | |
| BC055862.1 | - | 5 - 6 | 396 | 3492090 | 67 | |
| BC069899.1 | - | 4 - 5 | 1118 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28975560 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28975561 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28975562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28975559 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P338600 | BC025854.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P338600 | BC055862.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P338600 | BC069899.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P338600 | CO039120.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P338600 | AK003891.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P338600 | BC025854.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P338600 | BC055862.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P338600 | BC069899.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P338600 | CO039120.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P338600 | AK003891.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926980 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308417 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35612 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116858 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P338600 | BC025854.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P338600 | BC055862.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P338600 | BC069899.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P338600 | CO039120.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P338600 | AK003891.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P338600 | BC025854.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P338600 | BC055862.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P338600 | BC069899.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P338600 | CO039120.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P338600 | AK003891.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975555 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28975558 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02673651 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116858 | - | - |
| Whole Mouse Genome G4122A | A_51_P338600 | BC025854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338600 | BC055862.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338600 | BC069899.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338600 | CO039120.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338600 | AK003891.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338600 | BC025854.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338600 | BC055862.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338600 | BC069899.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338600 | CO039120.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338600 | AK003891.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106439 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120083 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116858 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11000889 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11025662 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11000890 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11000886 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11025658 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11025669 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11025656 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11025667 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28975562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28975559 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28975561 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28975560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28975610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11099842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10937953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10474261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11000886 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10677863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11099846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10893812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11025659 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11099837 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10792805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11099828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11097140 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975548 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11025663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11099832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10656408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11025656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28975571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11099835 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404735 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404734 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404732 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404733 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416479_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404731 | - | - |
| Mouse Genome 430 2.0 Array | 1416479_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416479_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66154_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66154_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300002754.mm.1 | NM_025387.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002754.mm.1 | NM_025387.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002754.mm.1 | TC1300000463.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416479_a_at | - | - |
| Mouse Expression Set 430 | 1416479_PM_a_at | - | - |
| Murine Genome U74v2 Set | 96353_PM_at | - | - |
| Murine Genome U74v2 Set | 96353_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416479_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416479_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416479_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416479_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 670647 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3940193 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 670647 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3940193 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000670647 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003940193 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2435206 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2472110 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2435206 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2472110 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003742.1_4-S | - | - |
| mouse-6 v1.1 (Illumina) | scl066154.6_169-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3940193 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 670647 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003742.1_4-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066154.6_169-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670647 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3940193 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2435206 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2472110 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2849680 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2472110 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2435206 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2849680 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066154.6_169-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003742.1_4-S | - | - |

Tmem14c
Genomic Map