Assay Details
| Gene Symbol: Smim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025471.2 | NP_079747.1 | 3 - 4 | 453 | 64 | |
| GenBank mRNA | AK005941.1 | - | 3 - 4 | 456 | 64 | |
| AK007303.1 | - | 2 - 3 | 211 | 64 | ||
| AK007660.1 | - | 2 - 3 | 231 | 64 | ||
| BC027508.1 | - | 2 - 3 | 201 | 1494969 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2454004 | AK005941.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2454004 | AK007303.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2454004 | NM_025471.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2454004 | AK007660.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2454004 | AK021236.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310576 | AK005941.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P310576 | NM_025471.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 808002 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88803 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71049 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179716 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310576 | AK005941.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P310576 | NM_025471.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2454004 | AK005941.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2454004 | AK007303.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2454004 | NM_025471.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2454004 | AK007660.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2454004 | AK021236.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22832498 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05199643 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179716 | - | - |
| Whole Mouse Genome G4122A | A_52_P534833 | AK005941.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P534833 | AK007303.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P534833 | NM_025471.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P534833 | AK007660.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P534833 | AK021236.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113148 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179716 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03705532 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03705536 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03705535 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03705539 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03705534 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03705541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22832549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03304250 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832522 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03929494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03929491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03929493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832514 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832496 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03705531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03816097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03705539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03304248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832473 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03862766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03929497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03689565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22832538 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10512006 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512007 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10512010 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430292_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10512005 | - | - |
| Mouse Genome 430 2.0 Array | 1430292_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430292_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66291_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66291_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400019415.mm.1 | NM_025471.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400019415.mm.1 | TC0400002391.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430292_a_at | - | - |
| Mouse Expression Set 430 | 1430292_PM_a_at | - | - |
| Murine Genome U74v2 Set | 170865_i_at | - | - |
| Murine Genome U74v2 Set | 170865_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430292_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1430292_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430292_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430292_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 742821 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 106200010 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6590746 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5390736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005390736 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006590746 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221983 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1246756 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246756 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221983 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2447708 | - | - |
| mouse-6 v1.1 (Illumina) | scl24463.1_27-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0002717.1_84-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0066291.2_269-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106200010 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5390736 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6590746 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002717.1_84-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066291.2_269-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5390736 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106200010 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590746 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221983 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2873180 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1246756 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221983 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2447708 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246756 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2873180 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0066291.2_269-S | - | - |

Smim8
Genomic Map