Assay Details
| Gene Symbol: Nosip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163684.1 | NP_001157156.1 | 4 - 5 | 362 | 63 | |
| NM_025533.3 | NP_079809.1 | 4 - 5 | 362 | 63 | ||
| GenBank mRNA | AK007962.1 | - | 4 - 5 | 311 | 63 | |
| AK010006.1 | - | 4 - 5 | 298 | 63 | ||
| BC089029.1 | - | 4 - 5 | 288 | 30624825 | 63 | |
| DV648837.1 | - | 4 - 5 | 362 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091032 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091030 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991931 | NM_025533.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991931 | AK005700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991931 | AK007962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991931 | AK010006.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991931 | AK076736.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40008 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854119 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877839 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484539 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646167 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 149155 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239365 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22784 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169963 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991931 | NM_025533.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991931 | AK005700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991931 | AK007962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991931 | AK010006.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991931 | AK076736.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25090984 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P169963 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06181848 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01507097 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01507102 | - | - |
| Whole Mouse Genome G4122A | A_52_P61811 | AK199797.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P61811 | AK219239.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P61811 | NM_025533.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P61811 | AK005700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P61811 | NM_001163684.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479177 | BC089029.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479177 | NM_025533.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479177 | AK007962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479177 | AK010006.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P479177 | AK076736.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134577 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P169963 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05892826 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091031 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091030 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25091032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25091043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25090980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06283363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06395355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06376775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06347637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06402747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06229296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06416438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06402739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06395357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06347639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06416435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06406724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06008158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06156539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06333415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06180057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25091007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06251084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25090973 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10552841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552839 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552833 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552837 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552835 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552838 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10552836 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418965_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431428_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10552832 | - | - |
| Mouse Genome 430 2.0 Array | 1418965_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431428_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1431428_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1418965_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66394_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66394_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700007065.mm.1 | NM_001163684.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007065.mm.1 | NM_001163684.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007065.mm.1 | NM_025533.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007065.mm.1 | NM_025533.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700007065.mm.1 | TC0700000827.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431428_a_at | - | - |
| Mouse Expression Set 430 | 1418965_at | - | - |
| Mouse Expression Set 430 | 1418965_PM_at | - | - |
| Mouse Expression Set 430 | 1431428_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431428_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431428_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418965_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418965_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418965_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418965_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431428_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431428_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 548998 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6380064 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6290671 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6380064 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6290671 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006380064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006290671 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225133 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2621588 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2621588 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225133 | - | - |
| mouse-6 v1.1 (Illumina) | scl00064.1_2-S | - | - |
| mouse-6 v1.1 (Illumina) | scl32695.9.13_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6290671 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6380064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00064.1_2-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32695.9.13_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6290671 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6380064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225133 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2621588 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2890743 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225133 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2890743 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2621588 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl32695.9.13_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00064.1_2-S | - | - |

Nosip
Genomic Map