Assay Details
| Gene Symbol: Nosip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163684.1 | NP_001157156.1 | 5 - 6 | 528 | 108 | |
| NM_025533.3 | NP_079809.1 | 5 - 6 | 528 | 108 | ||
| GenBank mRNA | AK007962.1 | - | 5 - 6 | 477 | 108 | |
| AK010006.1 | - | 5 - 6 | 464 | 108 | ||
| AK076736.1 | - | 5 - 6 | 453 | 108 | ||
| AK183200.1 | - | 1 - 2 | 109 | |||
| BC089029.1 | - | 5 - 6 | 454 | 30624825 | 108 | |
| DV648837.1 | - | 5 - 6 | 528 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nosip
Genomic Map