Assay Details
| Gene Symbol: Nosip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025533.3 | NP_079809.1 | 6 - 7 | 643 | 74 | |
| GenBank mRNA | AK010006.1 | - | 6 - 7 | 579 | 74 | |
| AK076736.1 | - | 6 - 7 | 568 | 74 | ||
| BC089029.1 | - | 6 - 7 | 569 | 30624825 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nosip
Genomic Map