Assay Details
| Gene Symbol: Nosip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163684.1 | NP_001157156.1 | 7 - 8 | 758 | 92 | |
| NM_025533.3 | NP_079809.1 | 7 - 8 | 833 | 92 | ||
| GenBank mRNA | AK007962.1 | - | 7 - 8 | 707 | 92 | |
| AK010006.1 | - | 7 - 8 | 769 | 92 | ||
| AK076736.1 | - | 7 - 8 | 758 | 92 | ||
| AK184378.1 | - | 1 - 2 | 63 | 92 | ||
| BC089029.1 | - | 7 - 8 | 759 | 30624825 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nosip
Genomic Map